Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083382.1 | NP_001076851.1 | 1 - 2 | 59 | 73 | |
GenBank mRNA | BC123773.1 | - | 1 - 2 | 59 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |