Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083526.1 | NP_001076995.1 | 7 - 8 | 573 | 158 | |
GenBank mRNA | BC114756.1 | - | 7 - 8 | 573 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |