Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083531.1 | NP_001077000.1 | 7 - 8 | 712 | 89 | |
GenBank mRNA | BC114766.1 | - | 7 - 8 | 712 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |