Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2IRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083745.1 | NP_001077214.1 | 13 - 14 | 1209 | 118 | |
GenBank mRNA | BC134477.1 | - | 13 - 14 | 1209 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |