Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097561.1 | NP_001091030.1 | 7 - 8 | 637 | 78 | |
GenBank mRNA | BC140489.1 | - | 7 - 8 | 637 | 78 | |
M16480.1 | - | 6 - 7 | 456 | 78 | ||
M16539.1 | - | 2 - 3 | 193 | 78 | ||
M36431.1 | - | 6 - 7 | 456 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |