Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098857.3 | NP_001092327.1 | 7 - 8 | 1033 | 109 | |
GenBank mRNA | AY214328.1 | - | 7 - 8 | 972 | 109 | |
BC142357.1 | - | 7 - 8 | 1034 | 109 | ||
BC151410.1 | - | 7 - 8 | 1034 | 109 | ||
DN513395.1 | - | 2 - 3 | 339 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |