Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174077.5 | NP_776502.1 | 2 - 3 | 329 | 59 | |
GenBank mRNA | BC149266.1 | - | 2 - 3 | 308 | 59 | |
DT856720.1 | - | 2 - 3 | 306 | 59 | ||
DT861549.1 | - | 2 - 3 | 299 | 59 | ||
DV837910.1 | - | 2 - 3 | 307 | 59 | ||
EV679018.1 | - | 2 - 3 | 309 | 59 | ||
L10325.1 | - | 2 - 3 | 254 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |