Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024484.2 | NP_001019655.2 | 3 - 4 | 296 | 71 | |
GenBank mRNA | BC102998.1 | - | 3 - 4 | 296 | 71 | |
BT021730.1 | - | 3 - 4 | 295 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |