Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034374.2 | NP_001029546.1 | 4 - 5 | 744 | 63 | |
GenBank mRNA | BC102764.1 | - | 4 - 5 | 758 | 63 | |
DV848567.1 | - | 4 - 5 | 562 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |