Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035271.4 | NP_001030348.2 | 3 - 4 | 1013 | 112 | |
GenBank mRNA | BC107527.1 | - | 3 - 4 | 1230 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |