Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097989.1 | NP_001091458.1 | 5 - 6 | 689 | 84 | |
GenBank mRNA | BC140659.1 | - | 5 - 6 | 689 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |