Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098379.1 | NP_001091849.1 | 2 - 3 | 252 | 79 | |
GenBank mRNA | EF560596.1 | - | 2 - 3 | 252 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |