Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098877.2 | NP_001092347.1 | 3 - 4 | 346 | 66 | |
GenBank mRNA | BC146028.1 | - | 3 - 4 | 348 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |