Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098890.2 | NP_001092360.1 | 3 - 4 | 381 | 78 | |
GenBank mRNA | BC142400.1 | - | 3 - 4 | 383 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |