Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098892.2 | NP_001092362.1 | 3 - 4 | 757 | 120 | |
GenBank mRNA | BC146026.1 | - | 3 - 4 | 758 | 120 | |
BC149415.1 | - | 5 - 6 | 695 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |