Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098912.1 | NP_001092382.1 | 9 - 10 | 851 | 66 | |
GenBank mRNA | BC142487.1 | - | 9 - 10 | 851 | 66 | |
BC151490.1 | - | 9 - 10 | 865 | 66 | ||
BC151824.1 | - | 9 - 10 | 851 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |