Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098944.1 | NP_001092414.1 | 4 - 5 | 1106 | 81 | |
GenBank mRNA | BC146247.1 | - | 4 - 5 | 1106 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |