Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAPLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098981.2 | NP_001092451.1 | 2 - 3 | 216 | 65 | |
GenBank mRNA | BC142409.1 | - | 2 - 3 | 217 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |