Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099377.1 | NP_001092847.1 | 1 - 2 | 420 | 120 | |
GenBank mRNA | BC146061.1 | - | 1 - 2 | 420 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |