Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100345.2 | NP_001093815.1 | 7 - 8 | 1378 | 101 | |
GenBank mRNA | BC148129.1 | - | 7 - 8 | 1381 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |