Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100359.3 | NP_001093829.1 | 6 - 7 | 1155 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |