Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100360.1 | NP_001093830.1 | 4 - 5 | 730 | 111 | |
GenBank mRNA | BC150014.1 | - | 4 - 5 | 730 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |