Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100380.1 | NP_001093850.1 | 4 - 5 | 2491 | 104 | |
GenBank mRNA | BC150096.1 | - | 4 - 5 | 2491 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |