Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100387.1 | NP_001093857.1 | 3 - 4 | 355 | 67 | |
GenBank mRNA | BC147923.1 | - | 3 - 4 | 355 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |