Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC90B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101863.1 | NP_001095333.1 | 8 - 9 | 708 | 88 | |
GenBank mRNA | BC149222.1 | - | 8 - 9 | 708 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |