Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101995.1 | NP_001095465.1 | 3 - 4 | 563 | 118 | |
GenBank mRNA | BC149081.1 | - | 3 - 4 | 563 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |