Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC18C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102034.1 | NP_001095504.1 | 7 - 8 | 946 | 79 | |
GenBank mRNA | BC149637.1 | - | 7 - 8 | 946 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |