Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102061.2 | NP_001095531.1 | 4 - 5 | 1182 | 79 | |
GenBank mRNA | BC150043.1 | - | 4 - 5 | 1184 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |