Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104982.1 | NP_001098452.1 | 1 | 228 | 98 | |
GenBank mRNA | BC150029.1 | - | 1 | 228 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |