Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109792.2 | NP_001103262.1 | 7 - 8 | 1175 | 86 | |
GenBank mRNA | AY265992.1 | - | 3 - 4 | 403 | 86 | |
BC153845.1 | - | 7 - 8 | 1177 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |