Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113299.2 | NP_001106770.1 | 2 - 3 | 302 | 68 | |
GenBank mRNA | BC116080.1 | - | 2 - 3 | 269 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |