Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113720.2 | NP_001107192.1 | 2 - 3 | 344 | 82 | |
GenBank mRNA | BC120474.1 | - | 2 - 3 | 345 | 82 | |
BC151417.1 | - | 2 - 3 | 293 | 82 | ||
DQ372929.1 | - | 2 - 3 | 345 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |