Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024565.2 | NP_001019736.2 | 3 - 4 | 254 | 93 | |
GenBank mRNA | BC111205.1 | - | 3 - 4 | 255 | 93 | |
BT021889.1 | - | 3 - 4 | 237 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |