Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076418.2 | NP_001069886.2 | 2 - 3 | 342 | 74 | |
GenBank mRNA | BC109547.1 | - | 2 - 3 | 245 | 74 | |
BT030599.2 | - | 2 - 3 | 341 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |