Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076902.2 | NP_001070370.2 | 1 - 2 | 88 | 67 | |
GenBank mRNA | EE341048.1 | - | 1 - 2 | 45 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |