Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098059.1 | NP_001091528.1 | 4 - 5 | 643 | 86 | |
GenBank mRNA | BT030596.2 | - | 4 - 5 | 641 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |