Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEMIS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098116.1 | NP_001091585.1 | 5 - 6 | 2037 | 100 | |
GenBank mRNA | BC140469.1 | - | 5 - 6 | 2037 | 100 | |
BC149610.1 | - | 5 - 6 | 1679 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |