Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098118.1 | NP_001091587.1 | 5 - 6 | 618 | 80 | |
GenBank mRNA | BC140603.1 | - | 5 - 6 | 618 | 80 | |
BC142259.1 | - | 5 - 6 | 715 | 80 | ||
BC142462.1 | - | 5 - 6 | 572 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |