Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYKK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099047.2 | NP_001092517.1 | 5 - 6 | 927 | 95 | |
GenBank mRNA | BC142244.1 | - | 5 - 6 | 928 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |