Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099141.1 | NP_001092611.1 | 2 - 3 | 607 | 53 | |
GenBank mRNA | BC134682.1 | - | 2 - 3 | 1329 | 53 | |
BC142129.1 | - | 2 - 3 | 607 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |