Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101126.2 | NP_001094596.1 | 2 - 3 | 688 | 88 | |
GenBank mRNA | BC151494.1 | - | 2 - 3 | 690 | 88 | |
BT029848.1 | - | 1 - 2 | 328 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |