Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101155.1 | NP_001094625.1 | 18 - 19 | 3600 | 76 | |
GenBank mRNA | BC148974.1 | - | 18 - 19 | 3600 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |