Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101170.2 | NP_001094640.2 | 3 - 4 | 425 | 79 | |
GenBank mRNA | BC126593.1 | - | 3 - 4 | 387 | 79 | |
DQ503408.1 | - | 3 - 4 | 425 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |