Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22H3orf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101260.1 | NP_001094730.1 | 3 - 4 | 668 | 80 | |
GenBank mRNA | BC147991.1 | - | 3 - 4 | 668 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |