Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANEAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102105.1 | NP_001095575.1 | 1 - 2 | 639 | 66 | |
GenBank mRNA | BC151266.1 | - | 1 - 2 | 639 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |