Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102111.1 | NP_001095581.1 | 3 - 4 | 593 | 84 | |
GenBank mRNA | BC149900.1 | - | 3 - 4 | 593 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |