Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102129.2 | NP_001095599.1 | 1 - 2 | 64 | 74 | |
GenBank mRNA | BC150053.1 | - | 1 - 2 | 65 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |