Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102167.1 | NP_001095637.1 | 4 - 5 | 1543 | 103 | |
GenBank mRNA | BC148040.1 | - | 4 - 5 | 1543 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |