Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102213.1 | NP_001095683.1 | 2 - 3 | 224 | 99 | |
GenBank mRNA | BC151356.1 | - | 2 - 3 | 224 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |