Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYRFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102323.1 | NP_001095793.1 | 21 - 22 | 2529 | 92 | |
GenBank mRNA | BC148934.1 | - | 21 - 22 | 2529 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |